Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100172.1 | NP_001093642.1 | 2 | 1606 | ||
GenBank mRNA | EF043041.1 | - | 2 | 1606 |