Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139114.3 | NP_620814.1 | 3, 4 | 384 | ||
GenBank mRNA | BC078724.1 | - | 2, 3 | 323 | 7125608 | |
FQ210520.1 | - | 3, 4 | 339 | |||
FQ217417.1 | - | 3, 4 | 340 | |||
FQ217514.1 | - | 3, 4 | 343 | |||
FQ218767.1 | - | 3, 4 | 339 | |||
FQ221301.1 | - | 3, 4 | 352 | |||
FQ221436.1 | - | 3, 4 | 341 | |||
FQ221541.1 | - | 3, 4 | 340 | |||
FQ221627.1 | - | 3, 4 | 343 | |||
FQ221775.1 | - | 3, 4 | 340 | |||
FQ221788.1 | - | 3, 4 | 339 | |||
FQ221816.1 | - | 3, 4 | 340 | |||
FQ222173.1 | - | 3, 4 | 340 | |||
FQ222227.1 | - | 3, 4 | 343 | |||
FQ222322.1 | - | 3, 4 | 339 | |||
FQ222450.1 | - | 3, 4 | 353 | |||
FQ222725.1 | - | 3, 4 | 340 | |||
FQ224390.1 | - | 3, 4 | 339 | |||
FQ227511.1 | - | 3, 4 | 354 | |||
FQ228660.1 | - | 2, 3 | 328 | |||
FQ230911.1 | - | 3, 4 | 343 | |||
X78167.1 | - | 2, 3 | 321 |
Gene Symbol: LOC102554047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770583.1 | XP_008768805.1 | 1 | 337 |
Gene Symbol: RGD1565131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248902.2 | XP_006248964.1 | 1 | 335 | ||
XM_008769031.1 | XP_008767253.1 | 1 | 335 |