Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005091.2 | NP_005082.1 | 3 | 808 | ||
GenBank mRNA | AF076483.1 | - | 3 | 575 | ||
AF242517.1 | - | 3 | 560 | |||
AY358936.1 | - | 3 | 556 | |||
BC096154.1 | - | 3 | 568 | 40003108 | ||
BC096155.3 | - | 3 | 568 | 40003109 | ||
BC096156.1 | - | 3 | 568 | 40003112 | ||
BC096157.1 | - | 3 | 568 | 40003114 | ||
BC101845.1 | - | 3 | 570 | 8069351 | ||
BC101847.1 | - | 3 | 570 | 8069353 |