Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039037.1 | NP_001034126.1 | 3, 4 | 337 | ||
XM_006229258.2 | XP_006229320.1 | 3, 4 | 356 | |||
XM_008759402.1 | XP_008757624.1 | 3, 4 | 335 | |||
GenBank mRNA | AY886529.1 | - | 3, 4 | 337 |