Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229462.2 | XP_006229524.1 | 5, 6 | 676 | ||
XM_006229463.2 | XP_006229525.1 | 5, 6 | 676 |