Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001281.1 | NP_001001281.1 | 1 | 420 |
Gene Symbol: Olr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001287.1 | NP_001001287.1 | 1 | 411 |
Set Membership: |
![]() |