Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014247.2 | NP_001014269.2 | 3 | 1318 | ||
XM_006231547.2 | XP_006231609.1 | 4 | 2368 | |||
XM_006231548.2 | XP_006231610.1 | 5 | 1569 | |||
XM_006231549.2 | XP_006231611.1 | 4 | 2797 | |||
XM_008760444.1 | XP_008758666.1 | 5 | 1560 | |||
GenBank mRNA | AY928801.1 | - | 3 | 1276 | ||
BC083580.1 | - | 5 | 1315 | 7129196 | ||
DQ176638.2 | - | 3 | 1276 |