Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127556.1 | NP_001121028.1 | 6 | 584 | ||
XM_006256320.2 | XP_006256382.1 | 7 | 678 | |||
XM_008772898.1 | XP_008771120.1 | 6 | 606 | |||
XM_008772899.1 | XP_008771121.1 | 5 | 350 | |||
GenBank mRNA | BC161883.1 | - | 6 | 440 | 7131276 |
Gene Symbol: LOC103694883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773854.1 | XP_008772076.1 | 25 |
Gene Symbol: LOC103694884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773855.1 | XP_008772077.1 | 5 | 401 | ||
XM_008773856.1 | XP_008772078.1 | 5 | 350 |