Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003969.3 | NP_003960.1 | 3 | 813 | ||
GenBank mRNA | AB012191.1 | - | 3 | 218 | ||
AF075599.1 | - | 3 | 221 | |||
BC058924.1 | - | 3 | 327 | 6162220 | ||
BT006754.1 | - | 3 | 218 |
Gene Symbol: UBE2MP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002837.1 | 1 | 367 | |||
GenBank mRNA | AW376368.1 | - | 1 | 169 |