Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037210.1 | NP_001032287.1 | 2 | 435 | ||
XM_008761552.1 | XP_008759774.1 | 2 | 1661 | |||
GenBank mRNA | BC100257.1 | - | 2 | 435 | 7442312 |