Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003979.3 | NP_003970.1 | 2 | 1514 | ||
GenBank mRNA | AF095448.1 | - | 2 | 963 | ||
AK001761.1 | - | 2 | 1117 | |||
AK122672.1 | - | 2 | 1754 | |||
AK172760.1 | - | 1 | 484 | |||
AK291171.1 | - | 2 | 949 | |||
BC003665.2 | - | 2 | 958 | 2988011 |
Set Membership: |
![]() |