Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108964.1 | NP_001102434.1 | 7, 8 | 657 | ||
XM_006235698.2 | XP_006235760.1 | 8, 9 | 817 | |||
XM_006235701.2 | XP_006235763.1 | 8, 9 | 817 | |||
XM_008762524.1 | XP_008760746.1 | 8, 9 | 817 | |||
XM_008762525.1 | XP_008760747.1 | 8, 9 | 1505 | |||
XM_008762527.1 | XP_008760749.1 | 8, 9 | 1051 |