Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267580.1 | NP_001254509.1 | 10, 11 | 1572 | ||
NM_003981.3 | NP_003972.1 | 11, 12 | 1695 | |||
NM_199413.2 | NP_955445.1 | 11, 12 | 1695 | |||
XM_005254987.2 | XP_005255044.1 | 11, 12 | 1603 | |||
XM_006720759.1 | XP_006720822.1 | 11, 12 | 1603 | |||
XM_006720760.1 | XP_006720823.1 | 11, 12 | 1603 | |||
XM_011522187.1 | XP_011520489.1 | 11, 12 | 1603 | |||
XM_011522188.2 | XP_011520490.1 | 11, 12 | 1603 | |||
XM_011522190.2 | XP_011520492.1 | 10, 11 | 1432 | |||
XM_011522191.2 | XP_011520493.1 | 11, 12 | 1603 | |||
XM_011522192.1 | XP_011520494.1 | 9, 10 | 1282 | |||
XM_017022714.1 | XP_016878203.1 | 10, 11 | 1477 | |||
XM_017022715.1 | XP_016878204.1 | 10, 11 | 1477 | |||
XM_017022716.1 | XP_016878205.1 | 10, 11 | 1432 | |||
XM_017022717.1 | XP_016878206.1 | 10, 11 | 1480 | |||
GenBank mRNA | AF044588.1 | - | 11, 12 | 1539 | ||
AK297117.1 | - | 10, 11 | 1446 | |||
AK312600.1 | - | 11, 12 | 1593 | |||
BC003138.1 | - | 11, 12 | 1563 | 2958690 | ||
BC005140.2 | - | 11, 12 | 1503 | 3504761 | ||
BG823292.1 | - | 1, 2 | 47 | |||
BX647317.1 | - | 2, 3 | 231 |