Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252065.1 | NP_001238994.1 | 9 | 1244 | ||
NM_001300773.1 | NP_001287702.1 | 9 | 1151 | |||
NM_004200.3 | NP_004191.2 | 8 | 1019 | |||
XM_005274383.4 | XP_005274440.1 | 12 | 1906 | |||
XM_005274384.2 | XP_005274441.1 | 11 | 1910 | |||
XM_005274385.4 | XP_005274442.1 | 11 | 1676 | |||
XM_005274387.4 | XP_005274444.1 | 6 | 1943 | |||
XM_005274390.4 | XP_005274447.1 | 5 | 982 | |||
XM_006718736.3 | XP_006718799.1 | 5 | 953 | |||
XM_011545335.2 | XP_011543637.1 | 12 | 1872 | |||
XM_011545336.2 | XP_011543638.1 | 11 | 1747 | |||
XM_011545337.2 | XP_011543639.1 | 11 | 1739 | |||
XM_011545338.2 | XP_011543640.1 | 11 | 1644 | |||
XM_011545339.2 | XP_011543641.1 | 10 | 1371 | |||
XM_011545340.2 | XP_011543642.1 | 9 | 1237 | |||
XM_011545341.2 | XP_011543643.1 | 9 | 1141 | |||
XM_011545343.2 | XP_011543645.1 | 8 | 1005 | |||
GenBank mRNA | AF038535.1 | - | 8 | 1012 | ||
BC125170.1 | - | 8 | 977 | 40121712 |