Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001387.1 | NP_001001387.1 | 1 | 843 |
Gene Symbol: Olr1051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001364.1 | NP_001001364.1 | 843 |