Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trhde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108991.1 | NP_001102461.1 | 2 | 1463 | ||
GenBank mRNA | X80535.1 | - | 1 | 522 |