Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134595.2 | NP_001128067.2 | 3 | 544 | ||
XM_006241491.2 | XP_006241553.1 | 3 | 2190 | |||
GenBank mRNA | FQ211772.1 | - | 3 | 480 |