Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004204.3 | NP_004195.2 | 5 | 1150 | ||
NM_148920.2 | NP_683721.1 | 5 | 1150 | |||
GenBank mRNA | AB003723.1 | - | 5 | 1114 | ||
AF030177.1 | - | 5 | 1083 | |||
AK300297.1 | - | 5 | 1117 | |||
AK314361.1 | - | 6 | 1179 | |||
BC006377.2 | - | 5 | 1100 | 4299132 |