Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004204.3 | NP_004195.2 | 6 | 1341 | ||
NM_148920.2 | NP_683721.1 | 6 | 1341 | |||
GenBank mRNA | AB003723.1 | - | 6 | 1305 | ||
AF030177.1 | - | 6 | 1274 | |||
AK300297.1 | - | 6 | 1308 | |||
AK314361.1 | - | 7 | 1370 | |||
BC006377.2 | - | 6 | 1291 | 4299132 |