Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005148.3 | NP_005139.1 | 4 | 640 | ||
NM_054035.2 | NP_473376.1 | 4 | 640 | |||
XM_017025289.1 | XP_016880778.1 | 5 | 665 | |||
GenBank mRNA | AB593014.1 | - | 4 | 643 | ||
AF028788.1 | - | 4 | 572 | |||
AF028789.1 | - | 4 | 572 | |||
AK124531.1 | - | 3 | 3446 | |||
AK292329.1 | - | 4 | 613 | |||
BC027176.1 | - | 4 | 614 | 4341426 | ||
BX641031.1 | - | 3 | 3369 | |||
BX647747.1 | - | 2 | 205 | |||
BX648530.1 | - | 3 | 1043 | |||
U40998.1 | - | 4 | 623 |