Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000637.1 | NP_001000637.1 | 1 | 123 |
Gene Symbol: Olr655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000342.1 | NP_001000342.1 | 123 |