Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004685.3 | NP_004676.3 | 2, 3 | 467 | ||
XM_011535306.1 | XP_011533608.1 | 2, 3 | 413 | |||
XM_011535307.1 | XP_011533609.1 | 2, 3 | 413 | |||
XM_017020845.1 | XP_016876334.1 | 2, 3 | 413 | |||
GenBank mRNA | AF406619.1 | - | 2, 3 | 148 | ||
AK314587.1 | - | 2, 3 | 332 | |||
AL832017.2 | - | 2, 3 | 390 | |||
BC040012.1 | - | 2, 3 | 339 | 5495564 |