Search Thermo Fisher Scientific
Gene Symbol: Olr1353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000750.1 | NP_001000750.1 | 1 | 664 | ||
XM_006245545.2 | XP_006245607.1 | 4 | 1154 | |||
XM_006245546.2 | XP_006245608.1 | 3 | 1006 |
Gene Symbol: Olr1350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000752.1 | NP_001000752.1 | 1 | 664 |