Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATP1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094673.1 | - | 1 | 1828 | ||
AK131061.1 | - | 7 | 950 | |||
AK293697.1 | - | 9 | 1065 | |||
AY946018.1 | - | 4 | 367 | |||
BC011835.2 | - | 7 | 942 | 3944210 | ||
U51478.1 | - | 7 | 819 |