Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004320.4 | NP_004311.1 | 1 | 269 | ||
NM_173201.3 | NP_775293.1 | 1 | 269 | |||
GenBank mRNA | AK291314.1 | - | 1 | 198 | ||
BC037354.1 | - | 1 | 197 | |||
BX537389.1 | - | 1 | 265 | |||
BX537784.1 | - | 1 | 267 | |||
BX647365.1 | - | 1 | 265 | |||
BX647366.1 | - | 1 | 261 | |||
BX647367.1 | - | 1 | 269 | |||
BX648544.1 | - | 1 | 266 | |||
BX649072.1 | - | 1 | 258 |