Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286075.1 | NP_001273004.1 | 17 | 2472 | ||
NM_004320.4 | NP_004311.1 | 19 | 2823 | |||
NM_173201.3 | NP_775293.1 | 19 | 2823 | |||
GenBank mRNA | AK092691.1 | - | 5 | 620 | ||
AK128456.1 | - | 17 | 2462 | |||
AK291314.1 | - | 19 | 2752 | |||
BX537389.1 | - | 19 | 2819 | |||
BX537784.1 | - | 19 | 2821 | |||
BX647365.1 | - | 19 | 2819 | |||
BX647366.1 | - | 19 | 2815 | |||
BX647368.1 | - | 12 | 1859 | |||
BX648544.1 | - | 19 | 2820 | |||
BX649072.1 | - | 19 | 2811 |