Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057749.2 | NP_477097.1 | 10 | 1029 | ||
XM_017013957.1 | XP_016869446.1 | 9 | 1354 | |||
XM_017013958.1 | XP_016869447.1 | 10 | 1171 | |||
XM_017013959.1 | XP_016869448.1 | 10 | 1273 | |||
GenBank mRNA | AF091433.1 | - | 9 | 962 | ||
AF102778.1 | - | 8 | 925 | |||
AF106690.1 | - | 8 | 925 | |||
AF112857.1 | - | 8 | 790 | |||
BC007015.1 | - | 8 | 764 | 3997513 | ||
BC020729.1 | - | 10 | 976 | 4774554 |