Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057749.2 | NP_477097.1 | 9 | 865 | ||
XM_011517366.2 | XP_011515668.1 | 8 | 1190 | |||
XM_017013957.1 | XP_016869446.1 | 8 | 1190 | |||
XM_017013958.1 | XP_016869447.1 | 9 | 1007 | |||
XM_017013959.1 | XP_016869448.1 | 9 | 1109 | |||
GenBank mRNA | AF091433.1 | - | 8 | 798 | ||
AF102778.1 | - | 7 | 761 | |||
AF106690.1 | - | 7 | 761 | |||
AF112857.1 | - | 7 | 626 | |||
BC007015.1 | - | 7 | 600 | 3997513 | ||
BC020729.1 | - | 9 | 812 | 4774554 |