Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214460.1 | NP_999625.1 | 1 | 881 |
Gene Symbol: Olr1720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001116.1 | NP_001001116.1 | 1 | 881 |