Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000903.1 | NP_001000903.1 | 1 | 234 |
Gene Symbol: Olr824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000905.1 | NP_001000905.1 | 1 | 234 |
Gene Symbol: Olr825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000904.1 | NP_001000904.1 | 1 | 234 |
Gene Symbol: Olr827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000902.1 | NP_001000902.1 | 1 | 234 |
Gene Symbol: Olr828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000991.1 | NP_001000991.1 | 1 | 234 |
Gene Symbol: Olr829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000901.1 | NP_001000901.1 | 1 | 234 | ||
XM_008763250.1 | XP_008761472.1 | 359 |