Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000926.1 | NP_001000926.1 | 331 |
Gene Symbol: Olr610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000336.1 | NP_001000336.1 | 1 | 331 |