Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004709.2 | NP_004700.1 | 1 | 315 | ||
GenBank mRNA | BC113600.1 | - | 1 | 184 | 8322652 | |
BC113604.1 | - | 1 | 184 | 8322656 | ||
Y08902.1 | - | 1 | 315 |
Gene Symbol: SLITRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262342.3 | XP_005262399.1 | 5 | 6891 | ||
XM_005262343.3 | XP_005262400.1 | 5 | 6901 | |||
XM_005262344.3 | XP_005262401.1 | 5 | 6879 | |||
XM_005262345.3 | XP_005262402.1 | 4 | 6742 |