Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001075.1 | NP_001001075.1 | 1 | 661 |
Gene Symbol: LOC103689955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765075.1 | XP_008763297.1 | 658 |
Gene Symbol: Olr952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000074.1 | NP_001000074.1 | 1 | 661 |