Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319224.1 | NP_001306153.1 | 12 | 2411 | ||
NM_003686.4 | NP_003677.4 | 11 | 2251 | |||
NM_006027.4 | NP_006018.4 | 11 | 2251 | |||
NM_130398.3 | NP_569082.2 | 13 | 2519 | |||
XM_006711840.2 | XP_006711903.1 | 13 | 2522 | |||
XM_011544321.1 | XP_011542623.1 | 12 | 2416 | |||
XM_011544322.1 | XP_011542624.1 | 12 | 2485 | |||
XM_011544323.2 | XP_011542625.1 | 13 | 2519 | |||
XM_011544324.2 | XP_011542626.1 | 12 | 2402 | |||
XM_011544325.2 | XP_011542627.1 | 5 | 1131 | |||
XM_017002793.1 | XP_016858282.1 | 11 | 2296 | |||
GenBank mRNA | AF042282.1 | - | 11 | 2155 | ||
AF060479.1 | - | 11 | 2146 | |||
AF084974.1 | - | 11 | 2146 | |||
AF091740.1 | - | 11 | 2148 | |||
AK291291.1 | - | 12 | 2399 | |||
AL080139.1 | - | 7 | 1417 | |||
BC007491.2 | - | 13 | 2467 | 2967734 |