Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001075.1 | NP_001001075.1 | 1 | 763 |
Gene Symbol: Olr952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000074.1 | NP_001000074.1 | 1 | 763 |