Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004070.2 | NP_001004070.1 | 2 | 152 | ||
NM_001165880.1 | NP_001159352.1 | 3 | 300 | |||
XM_006255977.2 | XP_006256039.1 | 3 | 1021 | |||
XM_006255978.2 | XP_006256040.1 | 4 | 907 | |||
XM_006255979.2 | XP_006256041.1 | 4 | 403 | |||
GenBank mRNA | BC127463.1 | - | 3 | 300 | 8362353 |