Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077651.1 | NP_001071119.1 | 5, 6 | 841 | ||
XM_006238591.2 | XP_006238653.1 | 5, 6 | 1002 | |||
XM_006238592.2 | XP_006238654.1 | 5, 6 | 1317 | |||
XM_006238593.2 | XP_006238655.1 | 5, 6 | 750 | |||
XM_006238594.2 | XP_006238656.1 | 5, 6 | 1026 | |||
XM_006238595.2 | XP_006238657.1 | 5, 6 | 1001 | |||
XM_008763893.1 | XP_008762115.1 | 5, 6 | 928 | |||
GenBank mRNA | BC158558.1 | - | 5, 6 | 852 | 8371379 | |
S83320.1 | - | 5, 6 | 841 |