Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF2 SIRNA Assays ›
Gene Symbol:
ARHGEF2
Gene Name:
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases:
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location:
Chr.1: 155946839 - 155991263 on Build GRCh38
UniGene ID:
Hs.743352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17547
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162383.1
NP_001155855.1
14
1888
NM_001162384.1
NP_001155856.1
14
1885
NM_004723.3
NP_004714.2
14
1813
XM_005245587.2
XP_005245644.1
14
1941
XM_005245588.2
XP_005245645.1
14
1938
XM_005245589.4
XP_005245646.1
14
2094
XM_005245590.3
XP_005245647.1
14
1816
XM_005245591.3
XP_005245648.1
15
1932
XM_005245592.2
XP_005245649.1
14
2085
XM_005245593.4
XP_005245650.1
14
1816
XM_005245594.2
XP_005245651.1
14
2802
XM_006711622.3
XP_006711685.1
14
3079
XM_006711623.3
XP_006711686.1
13
1815
XM_006711624.3
XP_006711687.1
13
1842
XM_006711625.3
XP_006711688.1
13
1748
XM_006711626.2
XP_006711689.1
13
2734
XM_011510136.2
XP_011508438.2
18
2497
XM_011510137.2
XP_011508439.2
18
2500
XM_011510139.2
XP_011508441.1
18
2500
XM_011510141.2
XP_011508443.1
14
2618
XM_011510143.2
XP_011508445.1
10
1364
XM_017002794.1
XP_016858283.1
18
2497
XM_017002795.1
XP_016858284.1
14
1832
XM_017002796.1
XP_016858285.1
14
3076
XM_017002797.1
XP_016858286.1
14
1816
XM_017002798.1
XP_016858287.1
13
3011
XM_017002799.1
XP_016858288.1
13
1812
GenBank mRNA
AB014551.3
-
18
2156
AF486838.1
-
14
1767
AL512715.1
-
14
1798
BC020567.1
-
14
1870
4157775
BT007407.1
-
13
1689
U72206.1
-
14
1875
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
cell morphogenesis
intracellular protein transport
actin filament organization
negative regulation of microtubule depolymerization
mitotic nuclear division
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
regulation of Rho protein signal transduction
intracellular signal transduction
regulation of cell proliferation
positive regulation of apoptotic process
positive regulation of GTPase activity
innate immune response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of neurogenesis
regulation of small GTPase mediated signal transduction
positive regulation of NF-kappaB transcription factor activity
cell division
negative regulation of necroptotic process
cellular response to muramyl dipeptide
cellular response to tumor necrosis factor
cellular hyperosmotic response
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
microtubule binding
transcription factor binding
zinc ion binding
Rho GTPase binding
Rac guanyl-nucleotide exchange factor activity
Rac GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0