Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002253.1 | NP_001002253.1 | 2 | 208 | ||
GenBank mRNA | BC089958.1 | - | 160 | 7312897 | ||
CB757317.1 | - | 2 | 192 | |||
FQ212114.1 | - | 1298 |