Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009499.1 | NP_001009499.1 | 271 | |||
XM_008763437.1 | XP_008761659.1 | 1 | 580 | |||
XM_008763438.1 | XP_008761660.1 | 2 | 538 | |||
GenBank mRNA | AY659932.1 | - | 271 |