Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009520.1 | NP_001009520.1 | 1 | 126 | ||
GenBank mRNA | AY510327.1 | - | 1 | 126 |
Gene Symbol: Vom1r72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009521.1 | NP_001009521.1 | 1 | 126 | ||
GenBank mRNA | AY510329.1 | - | 1 | 126 |