Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 6 | 1006 | ||
GenBank mRNA | AK299293.1 | - | 6 | 965 | ||
AK301461.1 | - | 5 | 774 | |||
AK310703.1 | - | 6 | 947 | |||
AK310938.1 | - | 4 | 506 | |||
AL389985.1 | - | 6 | 931 | |||
AL833263.1 | - | 5 | 729 | |||
BC033903.1 | - | 6 | 988 | |||
BC107880.1 | - | 6 | 987 |