Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 1 | 327 | ||
NM_001159739.2 | NP_001153211.1 | 2 | 297 | |||
NM_001287025.1 | NP_001273954.1 | 1 | 189 | |||
GenBank mRNA | BC100080.1 | - | 2 | 154 | 7324340 | |
CO566642.1 | - | 2 | 160 | |||
DY308874.1 | - | 2 | 221 | |||
DY573045.1 | - | 22 | ||||
FQ210706.1 | - | 1 | 156 | |||
FQ218526.1 | - | 2 | 170 | |||
FQ218726.1 | - | 2 | 164 | |||
FQ219557.1 | - | 2 | 164 | |||
FQ219704.1 | - | 2 | 164 | |||
FQ231428.1 | - | 2 | 168 | |||
S72506.1 | - | 1 | 175 | |||
S82820.1 | - | 1 | 327 | |||
X78847.1 | - | 1 | 175 |