Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 5 | 844 | ||
NM_001159739.2 | NP_001153211.1 | 6 | 814 | |||
NM_001287025.1 | NP_001273954.1 | 5 | 706 | |||
GenBank mRNA | DY573045.1 | - | 4 | 539 | ||
FQ210706.1 | - | 5 | 673 | |||
FQ218526.1 | - | 6 | 687 | |||
FQ218726.1 | - | 6 | 681 | |||
FQ219557.1 | - | 6 | 681 | |||
FQ219615.1 | - | 6 | 814 | |||
FQ219704.1 | - | 6 | 681 | |||
FQ231428.1 | - | 6 | 686 | |||
S72506.1 | - | 5 | 692 | |||
S82820.1 | - | 5 | 844 | |||
X78847.1 | - | 5 | 692 |