Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009975.1 | NP_001009975.1 | 1 | 923 |
Set Membership: |
G-protein coupled receptor |