Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135669.1 | NP_001129141.1 | 10 | 1405 | ||
NM_004736.3 | NP_004727.2 | 10 | 1405 | |||
GenBank mRNA | AF089744.1 | - | 10 | 1378 | ||
AF099082.1 | - | 10 | 1382 | |||
AF115389.1 | - | 10 | 1213 | |||
AK309750.1 | - | 10 | 1393 | |||
AL133058.1 | - | 7 | 907 | |||
AL137583.1 | - | 3 | 378 | |||
BC041142.1 | - | 10 | 1383 | 5582839 |
Set Membership: |
G-protein coupled receptor |