Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173374.1 | NP_001166845.1 | 3 | 463 | ||
GenBank mRNA | BC158882.1 | - | 3 | 398 | ||
CK359925.1 | - | 3 | 463 |