Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003574.5 | NP_003565.4 | 4 | 690 | ||
NM_194434.2 | NP_919415.2 | 4 | 690 | |||
XM_011525769.2 | XP_011524071.1 | 4 | 457 | |||
XM_017026079.1 | XP_016881568.1 | 4 | 466 | |||
GenBank mRNA | AF044670.1 | - | 4 | 394 | ||
AF057358.1 | - | 4 | 370 | |||
AF086627.1 | - | 4 | 370 | |||
AF154847.1 | - | 4 | 407 | |||
AK292948.1 | - | 4 | 418 | |||
AK315577.1 | - | 4 | 629 | |||
BC002992.1 | - | 4 | 604 | 2822547 | ||
BG488667.1 | - | 4 | 396 | |||
BG719406.1 | - | 4 | 668 | |||
BT019618.1 | - | 4 | 370 |