Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017480.1 | NP_001017480.1 | 1 | 268 | ||
GenBank mRNA | BC079340.1 | - | 1 | 268 | 7106610 |