Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284388.1 | NP_001271317.1 | 10 | 1424 | ||
NM_001284389.1 | NP_001271318.1 | 10 | 1397 | |||
NM_004741.4 | NP_004732.2 | 10 | 1394 | |||
XM_005270273.1 | XP_005270330.1 | 10 | 1401 | |||
GenBank mRNA | AK056755.1 | - | 9 | 1017 | ||
AK225101.1 | - | 10 | 1184 | |||
AK314363.1 | - | 10 | 1181 | |||
AY820769.1 | - | 10 | 1162 | |||
BC001883.1 | - | 10 | 1169 | 3533568 | ||
BC006769.2 | - | 6 | 679 | 2900024 | ||
BM473772.1 | - | 4 | 344 | |||
D21262.1 | - | 10 | 1185 | |||
Z34289.1 | - | 10 | 1213 |